missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Hyaluronan-binding protein 1

A DNA sequence encoding the Homo sapiens (Human) Hyaluronan-binding protein 2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7549-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16009330

  • £1735.00 / 1mg
Estimated Shipment: 29-08-2024
to see stock.

Description

Description

Specifications

Specifications

NP_001171131.1
Hyaluronan-binding protein 1
Research Use Only
HABP2
Recombinant Protein
E. coli
WTPDQYDYSY EDYNQEENTS STLTHAENPD WYYTEDQADP CQPNPCEHGG DCLVHGSTFT CSCLAPFSGN KCQKVQNTCK DNPCGRGQCL ITQSPPYYRC VCKHPYTGPS CSQVVPVCRP NPCQNGATCS RHKRRSKFTC ACPDQFKGKF CEIGSDDCYV GDGYSYRGKM NRTVNQHACL YWNSHLLLQE NYNMFMEDAE THGIGEHNFC RNPDADEKPW CFIKVTNDKV KWEYCDVSAC SAQDVAYPEE SPTEPSTKLP GFDSCGKTEI AERKIKR
Greater than 90% as determined by SDS-PAGE.
3026
1 mg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers