missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human IGFBP-rp1/IGFBP-7 Protein
Recombinant Protein
Brand: Novus Biologicals™ NBP2-35007-1mg
This item is not returnable.
View return policy
Specifications
Q16270, Q16270, Q16270 | |
PAGE | |
3490 | |
1 mg | |
Less than 0.1 EU/ug of IGFBP-rp1/IGFBP-7 as determined by LAL method. | |
IGFBP7 | |
Biologically Active Proteins | |
IGFBP-rp1/IGFBP-7 |
LYOPH | |
Lyophilized from a 0.2 um filtered concentrated solution in 20 mM Tris-HCl, pH 8.6, 150 mM NaCl. | |
26.4 kDa | |
Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. | |
angiomodulin, FSTL2, IBP-7, IGFBP-7PGI2-stimulating factor, IGFBP-7v, IGFBP-rP1, IGFBPRP1, insulin-like growth factor binding protein 7, insulin-like growth factor-binding protein 7, MAC25 protein, MAC25IGF-binding protein 7, Prostacyclin-stimulating factor, PSFAGM, TAF, Tumor-derived adhesion factor | |
Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0mg/mL. Apportion stock solutions into working aliquots and store at <-20°C. | |
>95% by SDS-PAGE and HPLC |