missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IHH Protein
A DNA sequence encoding the Homo sapiens (Human) IHH, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7597-ec-10ug
This item is not returnable.
View return policy
Specifications
NP_002172.2 | |
IHH Protein | |
Research Use Only | |
IHH | |
Recombinant Protein | |
E. coli | |
CFPAGAQVRL ESGARVALSA VRPGDRVLAM GEDGSPTFSD VLIFLDREPH RLRAFQVIET QDPPRRLALT PAHLLFTADN HTEPAARFRA TFASHVQPGQ YVLVAGVPGL QPARVAAVST HVALGAYAPL TKHGTLVVED VVASCFAAVA DHHLAQLAFW PLRLFHSLAW GSWTPGEGVH WYPQLLYRLG RLLLEEGSFH PLGMSGAGS | |
Greater than 90% as determined by SDS-PAGE. |
3549 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |