missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IHH Protein
A DNA sequence encoding the Homo sapiens (Human) IHH, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_002172.2 |
---|---|
Gene ID (Entrez) | 3549 |
Name | IHH Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16039650
|
enQuireBio™
QP7597-ec-10ug |
10 μg | |||||||
16079650
|
enQuireBio™
QP7597-ec-50ug |
50 μg | |||||||
16029650
|
enQuireBio™
QP7597-ec-100ug |
100 μg |
£384.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16059650
|
enQuireBio™
QP7597-ec-200ug |
200 μg | |||||||
16069650
|
enQuireBio™
QP7597-ec-500ug |
500 μg | |||||||
16049650
|
enQuireBio™
QP7597-ec-1mg |
1 mg |
£1735.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_002172.2 | |
IHH Protein | |
Not determined. | |
Full Length | |
Human | |
Tris-based buffer, 50% glycerol |
3549 | |
Research Use Only | |
IHH | |
Recombinant Protein | |
CFPAGAQVRL ESGARVALSA VRPGDRVLAM GEDGSPTFSD VLIFLDREPH RLRAFQVIET QDPPRRLALT PAHLLFTADN HTEPAARFRA TFASHVQPGQ YVLVAGVPGL QPARVAAVST HVALGAYAPL TKHGTLVVED VVASCFAAVA DHHLAQLAFW PLRLFHSLAW GSWTPGEGVH WYPQLLYRLG RLLLEEGSFH PLGMSGAGS |