missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human IL-20 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£292.00
Specifications
Gene ID (Entrez) | 50604 |
---|---|
Molecular Weight (g/mol) | 17.6 kDa |
Name | IL-20 |
Quantity | 25 μg |
Source | E. coli-derived human IL-20 protein Leu25-Glu176,with an N-terminal Met |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18168550
|
R&D Systems™
1102-IL-025 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
£292.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18116330
|
R&D Systems™
1102-IL-025/CF |
Lyophilized from a 0.2μm filtered solution in PBS. |
£292.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Recombinant Human IL-20 Protein is derived from E. coli. The Recombinant Human IL-20 Protein has been validated for the following applications: Bioactivity.Specifications
50604 | |
IL-20 | |
E. coli-derived human IL-20 protein Leu25-Glu176,with an N-terminal Met | |
Unconjugated | |
>97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
17.6 kDa | |
25 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |