missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human IL-28A/IFN-lambda 2 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£292.00
Specifications
Gene ID (Entrez) | 282616 |
---|---|
Molecular Weight (g/mol) | 20.8 kDa |
Name | IL-28A/IFN-lambda 2 |
Quantity | 25 μg |
Source | Mouse myeloma cell line,NS0-derived human IL-28A/IFN-lambda 2 protein, (N-terminus) Human IL-28A Val26-Val200 Accession &Num; NP_742150 DI 6-His tag (C-terminus) |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18196300
|
R&D Systems™
1587-IL-025/CF |
Lyophilized from a 0.2μm filtered solution in PBS. |
£292.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18156210
|
R&D Systems™
1587-IL-025 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
£292.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Recombinant Human IL-28A/IFN-lambda 2 Protein is derived from NS0. The Recombinant Human IL-28A/IFN-lambda 2 Protein has been validated for the following applications: Bioactivity.Specifications
282616 | |
IL-28A/IFN-lambda 2 | |
Mouse myeloma cell line,NS0-derived human IL-28A/IFN-lambda 2 protein, (N-terminus) Human IL-28A Val26-Val200 Accession &Num; NP_742150 DI 6-His tag (C-terminus) | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
20.8 kDa | |
25 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title