missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human IL-36 alpha/IL-1F6 (aa 6-158) Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand: R&D Systems™ 6995-IL-010/CF
222.89 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
The Recombinant Human IL-36 alpha/IL-1F6 (aa 6-158) Protein is derived from E. coli. The Recombinant Human IL-36 alpha/IL-1F6 (aa 6-158) Protein has been validated for the following applications: Bioactivity.Specifications
Lyophilized from a 0.2 μm filtered solution in Tris, NaCl, TCEP, EDTA and Tween™ 20 with Trehalose. | |
17 kDa | |
10 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
Reconstitute at 100μg/mL in Water. |
27179 | |
IL-36 alpha/IL-1F6 (aa 6-158) | |
E. coli-derived human IL-36 alpha/IL-1F6 protein Lys6-Phe158 | |
Monomer | |
Recombinant | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |