missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human IL-5 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand: R&D Systems™ 205-IL-025/CF
470.83 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
The Recombinant Human IL-5 Protein is derived from Sf 21 (stably transfected). The Recombinant Human IL-5 Protein has been validated for the following applications: Bioactivity.Specifications
Supplied as a 0.2μm filtered solution in PBS. | |
13.1 kDa (monomer) | |
25 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after opening. 3 months, -20 to -70° C under sterile conditions after opening. | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
3567 | |
IL-5 | |
Spodoptera frugiperda,Sf 21 (stably transfected)-derived human IL-5 protein Ile20-Ser134 | |
Disulfide-linked homodimer | |
Recombinant |