missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human IL-6 Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
100 μg
5 μg
500 μg
Unit Size:
100µg
1mg
500µg
5µg
Description
Assay #1:
- Fully biologically active when compared to standard. The ED50 as determined by a cell proliferation assay using IL-6-dependent murine 7TD1 cells is less than 0.1 ng/mL, corresponding to a specific activity of >1.0 x 107 IU/mg.
Assay #2:
- Fully biologically active when compared to standard. The ED50 as determined by a cell proliferation assay using IL-6-dependent murine T1165 cells is less than 0.8 ng/mL, corresponding to a specific activity of >1.25 x 106 IU/mg.
Specifications
Specifications
| Accession Number | P05231, P05231, P05231 |
| Concentration | LYOPH |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4. |
| Gene ID (Entrez) | 3569 |
| Molecular Weight (g/mol) | 20.8 kDa |
| Quantity | 500 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1.0 EU/ug of IL-6 as determined by LAL method. |
| Gene Alias | B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction