missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL10 / Interleukin-10 Protein
A DNA sequence encoding the Homo sapiens (Human) IL10 / Interleukin-10, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8553-ec-100ug
This item is not returnable.
View return policy
Specifications
NP_000563.1 | |
IL10 / Interleukin-10 Protein | |
Research Use Only | |
IL-10 | |
Recombinant Protein | |
E. coli | |
SPGQGTQSEN SCTHFPGNLP NMLRDLRDAF SRVKTFFQMK DQLDNLLLKE SLLEDFKGYL GCQALSEMIQ FYLEEVMPQA ENQDPDIKAH VNSLGENLKT LRLRLRRCHR FLPCENKSKA VEQVKNAFNK LQEKGIYKAM SEFDIFINYI EAYMTMKIRN | |
Greater than 90% as determined by SDS-PAGE. |
3586 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |