missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL13RA2 Protein
A cDNA sequence encoding the IL13RA2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12406-HIS-5ug
This item is not returnable.
View return policy
Specifications
3598 | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
ADLDTEIKVN PPQDFEIVDP GYLGYLYLQW QPPLSLDHFK ECTVEYELKY RNIGSETWKT IITKNLHYKD GFDLNKGIEA KIHTLLPWQC TNGSEVQSSW AETTYWISPQ GIPETKVQDM DCVYYNWQYL LCSWKPGIGV LLDTNYNLFY WYEGLDHALQ CVDYIKADGQ NIGCRFPYLE ASDYKDFYIC VNGSSENKPI RSSYFTFQLQ NIVKPLPPVY LTFTRESSCE IKLKWSIPLG PIPARCFDYE IEIREDDTTL VTATVENETY TLKTTNETRQ LCFVVRSKVN IYCSDDGIWS EWSDKQCWEG EDLSKKTLLR LEPKSCDKTH TCPPCPAPEL LGGPSVFLFP PKPKDTLMIS RTPEVTCVVV DVSHEDPEVK FNWYVDGVEV HNAKTKPREE QYNSTYRVVS VLTVLHQDWL NGKEYKCKVS NKALPAPIEK TISKAKGQPR EPQVYTLPPS RDELTKNQVS LTCLVKGFYP SDIAVEWESN GQPENNYKTT PPVLDSDGSF FLYSKLTVDK SRWQQGNVFS CSVMHEALHNHYTQKSLSLS PGKHHHHHH | |
Greater than 85.0% as determined by SDS-PAGE. |
IL13RA2 Protein | |
Research Use Only | |
IL13RA2 | |
Human | |
His | |
IL 13RA2 protein solution (0.5 mg/ml) contains Phosphate buffered saline (pH 7.4) and 10% glycerol. |