missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL2 / Interleukin-2 Protein
Recombinant Protein
£476.00 - £810.00
Specifications
Accession Number | NP_000577.2 |
---|---|
Gene ID (Entrez) | 3558 |
Name | IL2 / Interleukin-2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15965769
|
enQuireBio™
QP5245-500ug |
500 μg |
£476.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15955769
|
enQuireBio™
QP5245-1mg |
1 mg |
£810.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_000577.2 | |
IL2 / Interleukin-2 Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity in the CTLL-2 cell proliferation assay. | |
Human | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |
3558 | |
Research Use Only | |
IL2 | |
Recombinant Protein | |
MAPTSSSTKK TQLQLEHLLL DLQMILNGIN NYKNPKLTRM LTFKFYMPKK ATELKHLQCL EEELKPLEEV LNLAQSKNFH LRPRDLISNI NVIVLELKGS ETTFMCEYAD ETATIVEFLN RWITFCQSII STLT |