missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL28B Protein
A cDNA sequence encoding the IL28B was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12417-2ug
This item is not returnable.
View return policy
Specifications
282617 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
VPVARLRGAL PDARGCHIAQ FKSLSPQELQ AFKRAKDALE ESLLLKDCKC RSRLFPRTWD LRQLQVRERP VALEAELALT LKVLEATADT DPALGDVLDQ PLHTLHHILS QLRACIQPQP TAGPRTRGRL HHWLHRLQEA PKKESPGCLE ASVTFNLFRL LTRDLNCVAS GDLCVHHHHHH | |
Greater than 85.0% as determined by SDS-PAGE. |
IL28B Protein | |
Research Use Only | |
IFNL3 | |
Human | |
His | |
IL 28B protein solution (0.25 mg/ml) contains Phosphate buffered saline (pH 7.4), 20% glycerol, 1mM EDTA and 0.1mM PMSF. |