missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL4 / Interleukin-4 Protein

A DNA sequence encoding the Homo sapiens (Human) IL4 / Interleukin-4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8557-ec-200ug

 View more versions of this product

Product Code. 16005271

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_000580.1
IL4 / Interleukin-4 Protein
Research Use Only
IL4
Recombinant Protein
E. coli
HKCDITLQEI IKTLNSLTEQ KTLCTELTVT DIFAASKNTT EKETFCRAAT VLRQFYSHHE KDTRCLGATA QQFHRHKQLI RFLKRLDRNL WGLAGLNSCP VKEANQSTLE NFLERLKTIM REKYSKCSS
Greater than 90% as determined by SDS-PAGE.
3565
200 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers