missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL7 / Interleukin 7 Protein

A DNA sequence encoding the Homo sapiens(Human) IL7 / Interleukin 7 Protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9979-iv-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16079131

  • £1240.00 / 100µg
Estimated Shipment: 05-09-2024
to see stock.

Description

Description

Specifications

Specifications

NP_000871.1
IL7 / Interleukin 7 Protein
Research Use Only
IL7
Recombinant Protein
E. coli
DCDIEGKDGK QYESVLMVSI DQLLDSMKEI GSNCLNNEFN FFKRHICDAN KEGMFLFRAA RKLRQFLKMN STGDFDLHLL KVSEGTTILL NCTGQVKGRK PAALGEAQPT KSLEENKSLK EQKKLNDLCF LKRLLQEIKT CWNKILMGTK EH
Greater than 85% as determined by SDS-PAGE.
3574
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers