missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Integrin alpha 2 beta 1 Protein

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity

Brand:  R&D Systems™ 5698-A2-050

Product Code. 18127580

  • £445.00 / 50µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Measured by its binding ability in a functional ELISA. When bovine collagen-II is coated at 10 μg/mL, Recombinant Human Integrin alpha 2 beta 1 binds with an apparent Kd <1nM.
Specifications

Specifications

NP_002194 (Integrin alpha 2) & P05556 (Integrin beta 1)
Lyophilized from a 0.2 μm filtered solution in PBS.
M.W. (Observed): 140-150 kDa (alpha 2) & 120-130 kDa (beta 1), reducing conditions; M.W. (theoretical): 126 kDa (alpha 2) & 83.3 kDa (beta 1)
50 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
ITGA2
Recombinant
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Binding Activity
3673
Integrin alpha 2 beta 1
Chinese Hamster Ovary cell line, CHO-derived human Integrin alpha 2 beta 1 protein Human Integrin alpha 2 (Tyr30-Thr1132) (N-terminus) Accession # NP_002194 HPGGGSGGGS Human c-Jun (Arg276-His315) Human Integrin beta 1 (Gln21-Asp728) (N-terminus) Accession # P05556 GGGSGGGS Human c-Fos (Leu161-Ala199) (C-terminus)
<0.10 EU / 1 μg of the protein by the LAL method.
Unconjugated
Reconstitute at 200 μg/mL in PBS.
Integrin alpha 2 beta 1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.