missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Kallikrein 11 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity
Brand: R&D Systems™ 1595-SE-010
310.14 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
The Recombinant Human Kallikrein 11 Protein is derived from NS0. The Recombinant Human Kallikrein 11 Protein has been validated for the following applications: Enzyme Activity.Specifications
Supplied as a 0.2 μm filtered solution in Tris, NaCl and CaCl 2 . | |
27 kDa | |
10 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
11012 | |
Kallikrein 11 | |
Mouse myeloma cell line,NS0-derived human Kallikrein 11 protein Glu19-Asn250,with an N-terminal signal peptide and a C-terminal 10-His tag | |
Pro form | |
Recombinant |