missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Kallikrein 12 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity
Brand: R&D Systems™ 3095-SE-010
344.70 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
The Recombinant Human Kallikrein 12 Protein is derived from NS0. The Recombinant Human Kallikrein 12 Protein has been validated for the following applications: Enzyme Activity.Specifications
Supplied as a 0.2μm filtered solution in MES, NaCl and Glycerol. | |
26 kDa | |
10 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
43849 | |
Kallikrein 12 | |
Mouse myeloma cell line,NS0-derived human Kallikrein 12 protein Ala18-Asn248,with a C-terminal 10-His tag | |
Pro form | |
Recombinant |