missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Kallikrein 8/Neuropsin Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

Brand:  R&D Systems™ 2025-SE-010

310.14 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 18126510

  • £405.00 / 10µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The Recombinant Human Kallikrein 8/Neuropsin Protein is derived from NS0. The Recombinant Human Kallikrein 8/Neuropsin Protein has been validated for the following applications: Enzyme Activity.
Specifications

Specifications

Supplied as a 0.2μm filtered solution in Tris and NaCl.
Observed Molecular Wt.: 37-39 kDa, reducing conditions
10 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening.
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
11202
Kallikrein 8/Neuropsin
Mouse myeloma cell line,NS0-derived human Kallikrein 8/Neuropsin protein Gln29-Gly260,with a C-terminal 10-His tag
Pro form
Recombinant
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers