missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human KCTD4 Protein
A cDNA sequence encoding the KCTD4 was constructed and used to recombinantly synthesize the protein.
£99.15 - £1320.00
Specifications
Gene ID (Entrez) | 386618 |
---|---|
Name | KCTD4 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | KCTD4 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15950559
|
enQuireBio™
QP12481-2ug |
2 μg |
£99.15
2µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15940559
|
enQuireBio™
QP12481-10ug |
10 μg |
£155.00
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15930559
|
enQuireBio™
QP12481-100ug |
100 μg |
£1320.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
386618 | |
Research Use Only | |
KCTD4 | |
Human | |
His | |
KCTD4 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 40% glycerol and 1mM DTT. |
KCTD4 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMERKINR REKEKEYEGK HNSLEDTDQG KNCKSTLMTL NVGGYLYITQ KQTLTKYPDT FLEGIVNGKI LCPFDADGHY FIDRDGLLFR HVLNFLRNGE LLLPEGFREN QLLAQEAEFF QLKGLAEEVK SRWEKEQLTP RETTFLEITD NHDRSQGLRI FCNAPDFISK IKSRIVLVSK SRLDGFPEEF SISSNIIQFK YFIKSENGTR LVLKEDNTFV CTLETLKFEA IMMALKCGFR LLTSLDCSKG SIVHSDALHF IK | |
Greater than 85% as determined by SDS-PAGE. |