missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Kinetochore-associated protein DSN1 homolog Protein
A DNA sequence encoding the Homo sapiens (Human) Kinetochore-associated protein DSN1 homolog, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8173-ec-50ug
This item is not returnable.
View return policy
Specifications
NP_001138787.1 | |
Kinetochore-associated protein DSN1 homolog Protein | |
Research Use Only | |
DSN1 | |
Recombinant Protein | |
E. coli | |
MTSVTRSEII DEKGPVMSKT HDHQLESSLS PVEVFAKTSA SLEMNQGVSE ERIHLGSSPK KGGNCDLSHQ ERLQSKSLHL SPQEQSASYQ DRRQSWRRAS MKETNRRKSL HPIHQGITEL SRSISVDLAE SKRLGCLLLS SFQFSIQKLE PFLRDTKGFS LESFRAKASS LSEELKHFAD GLETDGTLQK CFEDSNGKAS DFSLEASVAE MKEYITKFSL ERQTWDQLLL HYQQEAKEIL SRGSTEAKIT EVKVEPMTYL GSSQNEVLNT KPDYQKILQN QSKVFDCMEL VMDELQGSVK QLQAFMDEST QCFQKVSVQL GKRSMQQLDP SPARKLLKLQ LQNPPAIHGS GSGSCQ | |
Greater than 80% as determined by SDS-PAGE. |
79980 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |