missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Kita-kyushu lung cancer antigen 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Kita-kyushu lung cancer antigen 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7682-ec-10ug
This item is not returnable.
View return policy
Specifications
NP_001017978.1 | |
Kita-kyushu lung cancer antigen 1 Protein | |
Research Use Only | |
CT83 | |
Recombinant Protein | |
E. coli | |
MNFYLLLASS ILCALIVFWK YRRFQRNTGE MSSNSTALAL VRPSSSGLIN SNTDNNLAVY DLSRDILNNF PHSIARQKRI LVNLSMVENK LVELEHTLLS KGFRGASPHR KST | |
Greater than 80% as determined by SDS-PAGE. |
203413 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-B2M | |
Tris-based buffer, 50% glycerol |