missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human LBP Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£343.00
Specifications
Gene ID (Entrez) | 3929 |
---|---|
Name | LBP |
Quantity | 25 μg |
Source | Mouse myeloma cell line,NS0-derived human LBP protein Ala26-Val481 |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18320398
|
R&D Systems™
870-LP-025/CF |
Lyophilized from a 0.2μm filtered solution in Tris, NaCl and EDTA. |
£343.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18106950
|
R&D Systems™
870-LP-025 |
Lyophilized from a 0.2μm filtered solution in Tris, NaCl and EDTA with BSA as a carrier protein. |
£343.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Recombinant Human LBP Protein is derived from NS0. The Recombinant Human LBP Protein has been validated for the following applications: Bioactivity.Specifications
3929 | |
25 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
LBP | |
Mouse myeloma cell line,NS0-derived human LBP protein Ala26-Val481 | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title