missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Legumain/Asparaginyl Endopeptidase
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity
Brand: R&D Systems™ 2199-CY-010
325.70 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
Measured by its ability to cleave the fluorogenic peptide substrate, N-carbobenzyloxy-Ala-Ala-Asn-7-amido-4-methylcoumarin (Z-AAN-AMC). The specific activity is >250 pmol/min/μg, as measured under the described conditions.Specifications
AAH03061.1 | |
Supplied as a 0.2 μm filtered solution in Tris, NaCl and Glycerol. | |
M.W. (Observed): 60 kDa, reducing conditions; M.W. (theoretical): 49 kDa | |
10 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. | |
<1.0 EU / 1 μg of the protein by the LAL method. | |
Unconjugated | |
>95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Enzyme Activity | |
5641 | |
Legumain/Asparaginyl Endopeptidase | |
Mouse myeloma cell line, NS0-derived human Legumain/Asparaginyl Endopeptidase protein Ile18-Tyr433, with an N-terminal 7-His tag | |
Pro form | |
LGMN | |
Recombinant | |
Legumain/Asparaginyl Endopeptidase |