missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human LHPP Protein

A DNA sequence encoding the Homo sapiens (Human) LHPP, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8086-ye-100ug

 View more versions of this product

Product Code. 16082581

  • £560.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_001161352.1
LHPP Protein
Research Use Only
LHPP
Recombinant Protein
Yeast
MAPWGKRLAG VRGVLLDISG VLYDSGAGGG TAIAGSVEAV ARLKRSRLKV RFCTNESQKS RAELVGQLQR LGFDISEQEV TAPAPAACQI LKEQGLRPYL LIHDGVRSEF DQIDTSNPNC VVIADAGESF SYQNMNNAFQ VLMELEKPVL ISLGKGRYYK ETSGLMLDVG PYMKALEYAC GIKAEVVGKP SPEFFKSALQ AIGVEAHQAV MIGDDIVGDV GGAQRCGMRA LQVRTGKFRP SDEHHPEVKA DGYVDNLAEA VDLLLQHADK
Greater than 90% as determined by SDS-PAGE.
64077
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers