missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LILRA5 Protein
A DNA sequence encoding the Homo sapiens (Human) LILRA5, was expressed in the hosts and tags indicated.
£183.90 - £2010.00
Specifications
Accession Number | NP_067073.1 |
---|---|
Gene ID (Entrez) | 353514 |
Name | LILRA5 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15945569
|
enQuireBio™
QP2841-ye-10ug |
10 μg | |||||||
15955559
|
enQuireBio™
QP2841-20ug |
20 μg |
£494.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15985569
|
enQuireBio™
QP2841-ye-50ug |
50 μg | |||||||
15935569
|
enQuireBio™
QP2841-ye-100ug |
100 μg |
£467.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15965569
|
enQuireBio™
QP2841-ye-200ug |
200 μg | |||||||
15975569
|
enQuireBio™
QP2841-ye-500ug |
500 μg | |||||||
15955569
|
enQuireBio™
QP2841-ye-1mg |
1 mg |
£2010.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_067073.1 | |
LILRA5 Protein | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
353514 | |
Research Use Only | |
LILRA5 | |
Recombinant Protein | |
Yeast | |
GNLSKATLWA EPGSVISRGN SVTIRCQGTL EAQEYRLVKE GSPEPWDTQN PLEPKNKARF SIPSMTEHHA GRYRCYYYSP AGWSEPSDPL ELVVTGFYNK PTLSALPSPV VTSGENVTLQ CGSRLRFDRF ILTEEGDHKL SWTLDSQLTP SGQFQALFPV GPVTPSHRWM LRCYGSRRHI LQVWSEPSDL LEIPVSGAAD NLSPSQNKSD SGTASHLQDY AVENLIR | |
Greater than 90% as determined by SDS-PAGE. |