Learn More
R&D Systems™ Recombinant Human LRP-1 Cluster IV Fc Chimera Protein
Brand:  R&D Systems™ 5395-L4B-050
Product Code. 18798896
  • £606.00 / 50µg
Explore more special offers
Description
Description
Measured by its binding ability in a functional ELISA. When Recombinant Human LRP-1 Cluster IV Fc Chimera (Catalog # 5395-L4B) is immobilized at 50 ng/mL (100 μL/well), Recombinant Human LRPAP (Catalog # 4296-LR ) binds with an ED50 of 0.0250-0.350 ng/mL.
Specifications
Specifications
Q07954.2
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
M.W. Observed: 100-120 kDa, under reducing conditions., M.W. Theoretical: 76.7 kDa
Chinese Hamster Ovary cell line, CHO-derived human LRP-1 Cluster IV protein Human LRP1-C4 (Ser3332-Asp3779) (N-terminus) Accession # Q07954.2 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
<0.10 EU per 1 μg of the protein by the LAL method.
Reconstitute at 500 μg/mL in PBS.
LRP-1 Cluster IV
Bioactivity
4035
50 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Unconjugated
>90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie® Blue Staining.