missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Lymphotoxin alpha2/beta1 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand: R&D Systems™ 679-TX-010
314.46 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
The Recombinant Human Lymphotoxin alpha2/beta1 Protein is derived from Sf 21 (baculovirus). The Recombinant Human Lymphotoxin alpha2/beta1 Protein has been validated for the following applications: Bioactivity.Specifications
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. | |
18 kDa (LT alpha), 18.5 kDa ( LTbeta) | |
10 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
4049 | |
Lymphotoxin α2/β1 | |
Spodoptera frugiperda,Sf 21 (baculovirus)-derived human Lymphotoxin protein Leu35-Leu205 (LT alpha ) & Leu54-Gly244 (LT beta ),both with an N-terminal Met & Thr41-Leu205 (LT alpha ) | |
Noncovalently-linked heterotrimer | |
Recombinant |