Learn More
R&D Systems™ Recombinant Human Lymphotoxin beta R (LTbR) Fc Chimera
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand:  R&D Systems™ 629-LR-100
Product Code. 18196720
  • £337.00 / 100µg
Explore more special offers
Description
Description
The Recombinant Human Lymphotoxin beta R (LTbR) Fc Chimera is derived from NS0. The Recombinant Human Lymphotoxin beta R (LTbR) Fc Chimera has been validated for the following applications: Bioactivity.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS.
49.6 kDa (monomer)
100 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
4055
Lymphotoxin beta R/TNFRSF3 Fc Chimera
Mouse myeloma cell line,NS0-derived human Lymphotoxin beta R/TNFRSF3 protein, (N-terminus) Human LT beta R (Ser28-Met227) & (Gln31-Met227) Accession &Num; NP_002333 DIEGRMD Human IgG1 (Pro100-Lys330) 6-His tag (C-terminus)
Disulfide-linked homodimer
Recombinant
Product Suggestions