missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human m7GpppN-mRNA hydrolase Protein
A DNA sequence encoding the Homo sapiens (Human) m7GpppN-mRNA hydrolase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7809-ye-500ug
This item is not returnable.
View return policy
Specifications
NP_001229306.1 | |
m7GpppN-mRNA hydrolase Protein | |
Research Use Only | |
DCP2 | |
Recombinant Protein | |
Yeast | |
METKRVEIPG SVLDDFCSRF ILHIPSEERD NAIRVCFQIE LAHWFYLDFY MQNTPGLPQC GIRDFAKAVF SHCPFLLPQG EDVEKVLDEW KEYKMGVPTY GAIILDETLE NVLLVQGYLA KSGWGFPKGK VNKEEAPHDC AAREVFEETG FDIKDYICKD DYIELRINDQ LARLYIIPGI PKDTKFNPKT RREIRNIEWF SIEKLPCHRN DMTPKSKLGL APNKFFMAIP FIRPLRDWLS RRFGDSSDSD NGFSSTGSTP AKPTVEKLSR TKFRHSQQLF PDGSPGDQWV KHRQPLQQKP YNNHSEMSDL LKGKKCEKKL HPRKLQDNFE TDAVYDLPSS SEDQLLEHAE GQPVACNGHC KFPFSSRAFL SFKFDHNAIM KILDL | |
Greater than 90% as determined by SDS-PAGE. |
167227 | |
500 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His | |
Tris-based buffer, 50% glycerol |