missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MAPK1, Active Protein
A cDNA sequence encoding the MAPK1, Active was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP14023-EC-100ug
This item is not returnable.
View return policy
Specifications
5594 | |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MAAAAAAGAG PEMVRGQVFD VGPRYTNLSY IGEGAYGMVC SAYDNVNKVR VAIKKISPFE HQTYCQRTLR EIKILLRFRH ENIIGINDII RAPTIEQMKD VYIVQDLMET DLYKLLKTQH LSNDHICYFL YQILRGLKYI HSANVLHRDL KPSNLLLNTT CDLKICDFGL ARVADPDHDH TGFLTEYVAT RWYRAPEIML NSKGYTKSID IWSVGCILAE MLSNRPIFPG KHYLDQLNHI LGILGSPSQE DLNCIINLKA RNYLLSLPHK NKVPWNRLFP NADSKALDLL DKMLTFNPHK RIEVEQALAH PYLEQYYDPS DEPIAEAPFK FDMELDDLPK EKLKELIFEE TARFQPGYRS | |
The purity is greater than 95%. |
MAPK1, Active Protein | |
Research Use Only | |
MAPK1 | |
Human | |
Untagged | |
MAPK1 protein solution is supplied in 50mM HEPES pH 7.5, 100mM NaCl, 5mM DTT and 20% glycerol. |