missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MARCKS-related Protein
A DNA sequence encoding the Homo sapiens (Human) MARCKS-related protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6346-ec-200ug
This item is not returnable.
View return policy
Specifications
NP_075385.1 | |
MARCKS-related Protein | |
Research Use Only | |
MARCKSL1 | |
Recombinant Protein | |
E. coli | |
MGSQSSKAPR GDVTAEEAAG ASPAKANGQE NGHVKSNGDL SPKGEGESPP VNGTDEAAGA TGDAIEPAPT SQGAEAKGEV PPKETPKKKK KFSFKKPFKL SGLSFKRNRK EGGGDSSASS PTEEEQEQGE IGACSDEGTA QEGKAAATPE SQEPQAKGAE ASAASEEEAG PQATEPSTPS GPESGPTPAS AEQNE | |
Greater than 80% as determined by SDS-PAGE. |
65108 | |
200 μg | |
Not determined. | |
Full Length of BC066915 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |