missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MED21 Protein
A cDNA sequence encoding the MED21 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12671-2ug
This item is not returnable.
View return policy
Specifications
MED21 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMADRLTQ LQDAVNSLAD QFCNAIGVLQ QCGPPASFNN IQTAINKDQP ANPTEEYAQL FAALIARTAK DIDVLIDSLP SEESTAALQA ASLYKLEEEN HEAATCLEDV VYRGDMLLEK IQSALADIAQ SQLKTRSGTH SQSLPDS | |
Greater than 90% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
MED21 protein solution (0.5 mg/ml) contains 20mM Tris-HCl buffer, (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |