missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Metallothionein-1G Protein

A DNA sequence encoding the Homo sapiens (Human) Metallothionein-1G, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8897-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16057061

  • £384.00 / 100µg
Estimated Shipment: 09-10-2024
to see stock.

Description

Description

Specifications

Specifications

NP_001288196.1
Metallothionein-1G Protein
Research Use Only
MT1G
Recombinant Protein
E. coli
MDPNCSCAAA GVSCTCASSC KCKECKCTSC KKSCCSCCPV GCAKCAQGCI CKGASEKCS
Greater than 90% as determined by SDS-PAGE.
4495
100 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers