missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Metallothionein-1G Protein
A DNA sequence encoding the Homo sapiens (Human) Metallothionein-1G, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8897-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001288196.1 | |
Metallothionein-1G Protein | |
Research Use Only | |
MT1G | |
Recombinant Protein | |
E. coli | |
MDPNCSCAAA GVSCTCASSC KCKECKCTSC KKSCCSCCPV GCAKCAQGCI CKGASEKCS | |
Greater than 90% as determined by SDS-PAGE. |
4495 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |