missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Methionine Aminopeptidase 1D Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand: R&D Systems™ 4714-ZN-020
This item is not returnable.
View return policy
Description
Measure by its ability to remove methionine from a fluorogenic peptide substrate H-Met-Gly-Pro-AMC (Catalog # ES017 ). The resulting GP-AMC is cleaved by Recombinant Human DPPIV/CD26 (Catalog # 9168-SE ). The specific activity is >30 pmol/min/μg, as measured under the described conditions.Specifications
Q6UB28 | |
Supplied as a 0.2 μm filtered solution in Tris, NaCl and Glycerol. | |
M.W. (Observed): 38 kDa, reducing conditions; M.W. (theoretical): 34 kDa | |
20 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. | |
METAP1D | |
Recombinant | |
Methionine Aminopeptidase 1 D/MAP1D |
Enzyme Activity | |
254042 | |
Methionine Aminopeptidase 1D/MAP1D | |
Spodoptera frugiperda, Sf 21 (baculovirus)-derived human Methionine Aminopeptidase 1 D/MAP1D protein Arg44-Ala335, with an N-terminal Met and a C-terminal 10-His tag | |
<1.0 EU / 1 μg of the protein by the LAL method. | |
Unconjugated | |
>95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction