missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MIP-3 beta / CCL19 Protein
Recombinant Protein
Brand: enQuireBio™ QP5258-500ug
This item is not returnable.
View return policy
Specifications
NP_006265.1 | |
MIP-3 beta / CCL19 Protein | |
Research Use Only | |
CCL19 | |
Recombinant Protein | |
E. coli | |
GTNDAEDCCL SVTQKPIPGY IVRNFHYLLI KDGCRVPAVV FTTLRGRQLC APPDQPWVER IIQRLQRTSA KMKRRSS | |
Greater than 95% as determined by SDS-PAGE. |
6363 | |
500 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Human MIP-3 beta / CCL19 induces Primary human T cell chemotaxis. | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |