missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Mitochondrial ornithine transporter 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Mitochondrial ornithine transporter 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8300-ec-500ug
This item is not returnable.
View return policy
Specifications
NP_055067.1 | |
Mitochondrial ornithine transporter 1 Protein | |
Research Use Only | |
SLC25A15 | |
Recombinant Protein | |
E. coli | |
MKSNPAIQAA IDLTAGAAGG TACVLTGQPF DTMKVKMQTF PDLYRGLTDC CLKTYSQVGF RGFYKGTSPA LIANIAENSV LFMCYGFCQQ VVRKVAGLDK QAKLSDLQNA AAGSFASAFA ALVLCPTELV KCRLQTMYEM ETSGKIAKSQ NTVWSVIKSI LRKDGPLGFY HGLSSTLLRE VPGYFFFFGG YELSRSFFAS GRSKDELGPV PLMLSGGVGG ICLWLAVYPV DCIKSRIQVL SMSGKQAGFI RTFINVVKNE GITALYSGLK PTMIRAFPAN GALFLAYEYS RKLMMNQLEA Y | |
Greater than 80% as determined by SDS-PAGE. |
10166 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |