missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MMP-2 Protein
A cDNA sequence encoding the MMP-2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10790-2ug
This item is not returnable.
View return policy
Specifications
MMP-2 Protein | |
Research Use Only | |
The activity was measured by its ability to cleave fluorogenic peptide substrate, Mca-PLGL-Dpa-AR-NH2 (RND,Catalog # ES001). , The specific activity is > 1,000 pmoles/min/microg. Recombinant Human MMP-2 protein pro form needs to be activated with p-aminophenylmercuric acetate (APMA). Activation Protocol:1. Dilute MMP2 to 100microg/ml in the Assay Buffer: 50mM Tris, 10mM CaCl2, 150mM NaCl, 0.05% (w/v) and Brij 35, pH 7.5. 2. Activate MMP2 by adding APMA to a final concentration of 1mM. (Sigma, Catalog # A9563) and 100mM stock in DMSO. 3. Incubate at 37C for 1 hour. | |
Human | |
His | |
, 10mM CaCl2, 150mM NaCl, 0.05% (w/v) and Brij 35, pH 7.5. 2. Activate MMP2 by adding APMA to a final concentration of 1mM. (Sigma, Catalog # A9563) and 100mM stock in DMSO. 3. Incubate at |
2 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
Human (HEK293) | |
MEALMARGAL TGPLRALCLL GCLLSHAAAA PSPIIKFPGD VAPKTDKELA VQYLNTFYGC PKESCNLFVL KDTLKKMQKF FGLPQTGDLD QNTIETMRKP RCGNPDVANY NFFPRKPKWD KNQITYRIIG YTPDLDPETV DDAFARAFQV WSDVTPLRFS RIHDGEADIM INFGRWEHGD GYPFDGKDGL LAHAFAPGTG VGGDSHFDDD ELWTLGEGQV VRVKYGNADG EYCKFPFLFN GKEYNSCTDT GRSDGFLWCS TTYNFEKDGK YGFCPHEALF TMGGNAEGQP CKFPFRFQGT SYDSCTTEGR TDGYRWCGTT EDYDRDKKYG FCPETAMSTV GGNSEGAPCV FPFTFLGNKY ESCTSAGRSD GKMWCATTAN YDDDRKWGFC PDQGYSLFLV AAHEFGHAMG LEHSQDPGAL MAPIYTYTKN FRLSQDDIKG IQELYGASPD IDLGTGPTPT LGPVTPEICK QDIVFDGIAQ IRGEIFFFKD RFIWRTVTPR DKPMGPLLVA TFWPELPEKI DAVYEAPQEE KAVFFAGNEY WIYSASTLER GYPKPLTSLG LPPDVQRVDA AFNWSKNKKT YIFAGDKFWR YNEVKKKMDP GFPKLIADAW NAIPDNLDAV VDLQGGGHSY FFKGAYYLKL ENQSLKSVKF GSIKSDWLGC | |
Greater than 95% as determined by SDS-PAGE. |