missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MOB3B Protein
A cDNA sequence encoding the MOB3B was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12713-2ug
This item is not returnable.
View return policy
Specifications
MOB3B Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSIALKQ VFNKDKTFRP KRKFEPGTQR FELHKRAQAS LNSGVDLKAA VQLPSGEDQN DWVAVHVVDF FNRINLIYGT ICEFCTERTC PVMSGGPKYE YRWQDDLKYK KPTALPAPQY MNLLMDWIEV QINNEEIFPT CVGVPFPKNF LQICKKILCR LFRVFVHVYI HHFDRVIVMG AEAHVNTCYK HFYYFVTEMN LIDRKELEPL KEMTSRMCH | |
Greater than 90% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The MOB3B solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 1mM DTT and 20% glycerol. |