missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human/Mouse/Rat BMP-2 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand: R&D Systems™ 355-BM-010
321.38 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
Measured by its ability to induce alkaline phosphatase production by ATDC5 mouse chondrogenic cells. Binnerts, M.E. et al. (2004) Biochem. Biophys. Res. Commun. 315(2):272. The ED50 for this effect is 40-200 ng/mL.Specifications
Lyophilized from a 0.2μm filtered solution in Glycine, Sucrose, Tween™ 80 and Glutamic Acid with BSA as a carrier protein. | |
13 kDa (monomer) | |
10 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
BDA2, BMP2, BMP-2, BMP-2A, BMP2ABone morphogenetic protein 2A, bone morphogenetic protein 2, SSFSC | |
Recombinant |
650 | |
BMP-2 | |
Chinese Hamster Ovary cell line,CHO-derived BMP-2 protein Gln283-Arg396 | |
Disulfide-linked homodimer | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |