missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human/Mouse Wnt-5a Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£432.00
Specifications
Gene ID (Entrez) | 7474 |
---|---|
Molecular Weight (g/mol) | 38 kDa |
Name | Wnt-5a |
Quantity | 10 μg |
Source | Chinese Hamster Ovary cell line,CHO-derived Wnt-5a protein Gln38-Lys380 |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18117580
|
R&D Systems™
645-WN-010 |
Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS with BSA as a carrier protein. |
£432.00
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18385597
|
R&D Systems™
645-WN-010/CF |
Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS. |
£432.00
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Recombinant Human/Mouse Wnt-5a Protein is derived from CHO. The Recombinant Human/Mouse Wnt-5a Protein has been validated for the following applications: Bioactivity.Specifications
7474 | |
Wnt-5a | |
Chinese Hamster Ovary cell line,CHO-derived Wnt-5a protein Gln38-Lys380 | |
Unconjugated | |
80%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
38 kDa | |
10 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title