missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MRAS Protein
A cDNA sequence encoding the MRAS was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12726-20ug
This item is not returnable.
View return policy
Specifications
MRAS Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MATSAVPSDN LPTYKLVVVG DGGVGKSALT IQFFQKIFVP DYDPTIEDSY LKHTEIDNQW AILDVLDTAG QEEFSAMREQ YMRTGDGFLI VYSVTDKASF EHVDRFHQLI LRVKDRESFP MILVANKVDL MHLRKITREQ GKEMATKHNI PYIETSAKDP PLNVDKAFHD LVRVIRQQIP EKSQKKKKKT KWRGDRATGT HKLQC | |
Greater than 90% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
MRAS protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 5mM DTT, 40% glycerol, 200mM NaCl and 2mM EDTA. |