missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Myosin light polypeptide 6 Protein
A DNA sequence encoding the Homo sapiens (Human) Myosin light polypeptide 6, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8495-ec-500ug
This item is not returnable.
View return policy
Specifications
NP_066299.2 | |
Myosin light polypeptide 6 Protein | |
Research Use Only | |
MYL6 | |
Recombinant Protein | |
E. coli | |
DFTEDQTAEF KEAFQLFDRT GDGKILYSQC GDVMRALGQN PTNAEVLKVL GNPKSDEMNV KVLDFEHFLP MLQTVAKNKD QGTYEDYVEG LRVFDKEGNG TVMGAEIRHV LVTLGEKMTE EEVEMLVAGH EDSNGCINYE AFVRHILSG | |
Greater than 90% as determined by SDS-PAGE. |
4637 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |