missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human N-alpha-acetyltransferase 50 Protein
A DNA sequence encoding the Homo sapiens (Human) N-alpha-acetyltransferase 50, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8171-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_079422.1 | |
N-alpha-acetyltransferase 50 Protein | |
Research Use Only | |
NAA50 | |
Recombinant Protein | |
E. coli | |
MKGSRIELGD VTPHNIKQLK RLNQVIFPVS YNDKFYKDVL EVGELAKLAY FNDIAVGAVC CRVDHSQNQK RLYIMTLGCL APYRRLGIGT KMLNHVLNIC EKDGTFDNIY LHVQISNESA IDFYRKFGFE IIETKKNYYK RIEPADAHVL QKNLKVPSGQ NADVQKTDN | |
Greater than 80% as determined by SDS-PAGE. |
80218 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |