missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NAPG / Gamma SNAP Protein
A DNA sequence encoding the Homo sapiens (Human) NAPG / Gamma SNAP, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8033-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_003817.1 | |
NAPG / Gamma SNAP Protein | |
Research Use Only | |
NAPG | |
Recombinant Protein | |
E. coli | |
MAAQKINEGL EHLAKAEKYL KTGFLKWKPD YDSAASEYGK AAVAFKNAKQ FEQAKDACLR EAVAHENNRA LFHAAKAYEQ AGMMLKEMQK LPEAVQLIEK ASMMYLENGT PDTAAMALER AGKLIENVDP EKAVQLYQQT ANVFENDERL RQAVELLGKA SRLLVRGRRF DEAALSIQKE KNIYKEIENY PTCYKKTIAQ VLVHLHRNDY VAAERCVRES YSIPGFNGSE DCAALEQLLE GYDQQDQDQV SDVCNSPLFK YMDNDYAKLG LSLVVPGGGI KKKSPATPQA KPDGVTATAA DEEEDEYSGG LC | |
Greater than 80% as determined by SDS-PAGE. |
8774 | |
200 μg | |
Not determined. | |
Full Length of BC001889 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |