missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NARS Protein
A cDNA sequence encoding the NARS was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12794-HIS-EC-10ug
This item is not returnable.
View return policy
Specifications
4677 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
GSSHHHHHH SSGLVPRGSH MGSMVLAELY VSDREGSDAT GDGTKEKPFK TGLKALMTVG KEPFPTIYVD SQKENERWNV ISKSQLKNIK KMWHREQMKS ESREKKEAED SLRREKNLEE AKKITIKNDP SLPEPKCVKI GALEGYRGQR VKVFGWVHRL RRQGKNLMFL VLRDGTGYLQ CVLADELCQC YNGVLLSTES SVAVYGMLNL TPKGKQAPGG HELSCDFWEL IGLAPAGGAD NLINEESDVD VQLNNRHMMI RGENMSKILK ARSMVTRCFR DHFFDRGYYE VTPPTLVQTQ VEGGATLFKL DYFGEEAFLT QSSQLYLETC LPALGDVFCI AQSYRAEQSR TRRHLAEYTH VEAECPFLTF DDLLNRLEDL VCDVVDRILK SPAGSIVHEL NPNFQPPKRP FKRMNYSDAI VWLKEHDVKK EDGTFYEFGE DIPEAPERLM TDTINEPILL CRFPVEIKSF YMQRCPEDSR LTESVDVLMP NVGEIVGGSM RIFDSEEILA GYKREGIDPT PYYWYTDQRK YGTCPHGGYG LGLERFLTWI LNRYHIRDVC LYPRFVQRCT P | |
Greater than 80.0% as determined by SDS-PAGE. |
NARS Protein | |
Research Use Only | |
NARS | |
Human | |
His | |
NARS protein solution (0.25 mg/ml) containing Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |