missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NDST1 Protein

A DNA sequence encoding the Homo sapiens (Human) NDST1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6409-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16003110

  • £1735.00 / 1mg
Estimated Shipment: 05-09-2024
to see stock.

Description

Description

Specifications

Specifications

NP_001287992.1
NDST1 Protein
Research Use Only
NDST1
Recombinant Protein
E. coli
GWKRGLEPSA DAPEPDCGDP PPVAPSRLLP LKPVQAATPS RTDPLVLVFV ESLYSQLGQE VVAILESSRF KYRTEIAPGK GDMPTLTDKG RGRFALIIYE NILKYVNLDA WNRELLDKYC VAYGVGIIGF FKANENSLLS AQLKGFPLFL HSNLGLKDCS INPKSPLLYV TRPSEVEKGV LPGEDWTVFQ SNHSTYEPVL LAKTRSSESI PHLGADAGLH AALHATVVQD LGLHDGIQRV LFGNNLNFWL HKLVFVDAVA FLTGKRLSLP LDRYILVDID DIFVGKEGTR MKVEDVKALF DTQNELRAHI PNFTFNLGYS GKFFHTGTNA EDAGDDLLLS YVKEFWWFPH MWSHMQPHLF HNQSVLAEQM ALNKKFAVEH GIPTDMGYAV APHHSGVYPV HVQLYEAWKQ VWSIRVTSTE EYPHLKPARY RRGFIHNGIM VLPRQTCGLF THTIFYNEYP GGSSELDKII NGGELFLTVL LNPVSAPQPM AAGEKGLLHS LSAADTGFLE PGKGGEA
Greater than 83.3% as determined by SDS-PAGE.
3340
1 mg
Not determined.
Full Length of Isoform 2
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers