missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NDUFV3 Protein

A cDNA sequence encoding the NDUFV3 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12822-5ug

 View more versions of this product

Product Code. 15961659

  • £99.15 / 5µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

4731
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSSAESGKS EKGQPQNSKK QSPPKKPAPV PAEPFDNTTY KNLQHHDYST YTFLDLNLEL SKFRMPQPSS GRESPRH
Greater than 85.0% as determined by SDS-PAGE.
NDUFV3 Protein
Research Use Only
NDUFV3
Human
His
The NDUFV3 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 2mM DTT, 2mM EDTA and 50% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers