missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Nectin-4 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand: R&D Systems™ 2659-N4-050
319.65 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
Measured by the ability of the immobilized protein to support the adhesion of NIH-3T3 mouse embryonic fibroblast cells. When 5 x 104 cells/well are added to recombinant human Nectin-4 coated plates (10 μg/mL with 100 μL/well), approximately >45% will adhere after 1 hour incubation at 37 °C. Optimal dilutions should be determined by each laboratory for each application.Specifications
NP_112178 | |
Lyophilized from a 0.2 μm filtered solution in PBS. | |
M.W. (Observed): 44 kDa, reducing conditions; M.W. (theoretical): 35.6 kDa | |
50 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. | |
<0.10 EU / 1 μg of the protein by the LAL method. | |
Unconjugated | |
Reconstitute at 200 μg/mL in PBS. | |
Nectin-4/PVRL4 |
Bioactivity | |
81607 | |
Nectin-4 | |
Mouse myeloma cell line, NS0-derived human Nectin-4 protein Gly27-Val351, with a C-terminal 6-His tag | |
Monomer | |
NECTIN4 | |
Recombinant | |
>90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. |